Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 422700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-270-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-270-0 Supplier Invitrogen™ Supplier No. 422700
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

MUPP1 Polyclonal Antibody for Western Blot, IHC (F)

MUPP1 (multi PDZ domain protein 1, MPDZ) was discovered as an interacting partner with the 5-HT2C receptor1, and was found to co-localize with 5-HT2A or 5-HT2C receptors in all regions of the mouse brain, including the choroid plexus. MUPP1 contains 13 PDZ domains but no apparent catalytic domain. MUPP1 is concentrated in tight junctions of polarized epithelial cells and binds to claudin-1 and junctional adhesion molecule (JAM). MUPP1 may therefore function as a multivalent scaffold protein that recruits various proteins to tight junctions. MUPP1 also interacts with claudin-8 and may play a role in tight junction barrier function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUPP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene MPDZ
Gene Accession No. Q8VBX6
Gene Alias AI225843; B930003D11Rik; DKFZp781P216; FLJ25909; FLJ34626; FLJ90240; HYC2; LOW QUALITY PROTEIN: multiple PDZ domain protein; MPDZ; Multi-PDZ domain protein 1; multiple PDZ domain crumbs cell polarity complex component; multiple PDZ domain protein; Mupp1
Gene Symbols MPDZ
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of mouse MUPP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17475
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.