Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MUPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542259 Supplier Invitrogen™ Supplier No. PA542259
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DEAINVLRQT PQRVRLTLYR DEAPYKEEEV CDTLTIELQK KPGKGLGLSI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

MUPP1 (multi PDZ domain protein 1, MPDZ) was discovered as an interacting partner with the 5-HT2C receptor1, and was found to co-localize with 5-HT2A or 5-HT2C receptors in all regions of the mouse brain, including the choroid plexus. MUPP1 contains 13 PDZ domains but no apparent catalytic domain. MUPP1 is concentrated in tight junctions of polarized epithelial cells and binds to claudin-1 and junctional adhesion molecule (JAM). MUPP1 may therefore function as a multivalent scaffold protein that recruits various proteins to tight junctions. MUPP1 also interacts with claudin-8 and may play a role in tight junction barrier function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUPP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MPDZ
Gene Accession No. O75970
Gene Alias AI225843; B930003D11Rik; DKFZp781P216; FLJ25909; FLJ34626; FLJ90240; HYC2; LOW QUALITY PROTEIN: multiple PDZ domain protein; MPDZ; Multi-PDZ domain protein 1; multiple PDZ domain crumbs cell polarity complex component; multiple PDZ domain protein; Mupp1
Gene Symbols MPDZ
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MPDZ.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8777
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.