Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Murashige and Skoog Media

Catalog No. ICN2623120 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 1 L
1 x 10 L
1 x 50 L
Product Type:
Murashige and Skoog Basal Medium
Murashige and Skoog Medium
Content And Storage:
0°C to 5°C
4°C
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Content And Storage
ICN2623120 10 x 1 L Murashige and Skoog Basal Medium 4°C
ICN2623220 10 x 1 L Murashige and Skoog Basal Medium 0°C to 5°C
ICN2623122 1 x 10 L Murashige and Skoog Basal Medium 4°C
ICN2610020 10 x 1 L Murashige and Skoog Medium 4°C
ICN2610024 1 x 50 L Murashige and Skoog Medium 4°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN2623120 Supplier MP Biomedicals™ Supplier No. 092623120
Add to Cart
Add to Cart

All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.

  • Each formulation is prepared according to the original specification, except where noted
  • Finished lots of media are plant cell tested
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Murashige and Skoog Basal Medium
Quantity 10 x 1 L
Format Solution
pH 3.75 to 4.75
Content And Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.