Learn More
Research Products International Corp Murashige & Skoog Modified Medium, NH4NO3 Free, 67.5 Grams of Powder, Makes 25 Liters of Solution
Supplier: Research Products International Corp M1060025.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962). Modified by eliminating ammonium nitrate. Concentrate: 2652.09 mg/L Hygroscopic
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.