Learn More
Bioworld Murashige and Skoog(MS) Medium, Modification No. 2, 50 L
Supplier:  Bioworld 306300734
Encompass_Preferred
Murashige & Skoog (MS) MediumContains Micronutrients and 3/4 Macronutrients as described by Murashige & Skoog (1967) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested
Catalog No. 50-254-705
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.