Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MURC Polyclonal Antibody, Invitrogen™

Catalog No. PIPA525555
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525555 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525555 Supplier Thermo Scientific Supplier No. PA525555

Rabbit Polyclonal Antibody

MURC induces RHOA activation and activates NPPA transcription and myofibrillar organization through the Rho/ROCK signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MURC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene CAVIN4
Gene Accession No. A2AMM0, Q5BKX8
Gene Alias 2310039E09Rik; Caveolae-associated protein 4; cavin 4; CAVIN4; cavin-4; Murc; muscle related coiled-coil protein; muscle-related coiled-coil protein; Muscle-restricted coiled-coil protein
Gene Symbols CAVIN4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 87-116 amino acids from the N-terminal region of human MURC
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347273, 68016
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.