Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murexide, ACS Reagent (Ammonium Purpurate), Reagents

Catalog No. 01337883
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-883 5 g
01-337-884 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-883 Supplier Reagents Holdings Llc Supplier No. C12519805C2
Add to Cart
Add to Cart

Murexide, ACS Reagent (Ammonium Purpurate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H8N6O6
Molecular Weight (g/mol) 284.19
InChI Key PCCZWUCAYILEMX-UHFFFAOYSA-O
Synonym Ammonium Purpurate, Color Index 56085
IUPAC Name ammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
SMILES [NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1

Specifications

Melting Point >300°C
Boiling Point 1500°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
pH Color Changes Pink (Excess Calcium) to Purple (Excess EDTA)
Color Red
Quantity 5 g
Physical Form Solid
Chemical Name or Material Murexide, ACS Reagent (Ammonium Purpurate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.