Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murexide Indicator, 0.15% (w/v) in Ethylene Glycol, Ricca Chemical
SDP

Catalog No. 522116 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5221-16 500 mL
5221-4 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5221-16 Supplier Ricca Chemical Company Supplier No. 522116
Add to Cart
Add to Cart

APHA for Calcium Testing

Specifications

Volume (Metric) 500 mL
Packaging Amber Glass Bottle
Quantity 500 mL
Test Method APHA for Calcium
Type Murexide Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.