Learn More
ECM Bioscience MuRF1 (C-terminal region) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience MP3401
Encompass_Preferred
This antibody was affinity purified using MuRF1 (C-terminal region) peptide (without carrier). The antibody detects 38 and 30 kDa* proteins corresponding to the apparent molecular mass of MuRF1 isoforms on SDS-PAGE immunoblots of mouse C2C12 cells, and detects a 38 kDa band in mouse heart and muscle tissue. The immunogen was MuRF1 (C-terminal) synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal half of human MuRF1. This peptide sequence is highly conserved in rat and mouse MuRF1, and has 50% homology to MuRF2 (TRIM-55). Accession #Q969Q1. Applications = WB, E, ICC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Tripartite motif 63,TRIM63, SMRZ, IRF, RNF28, Muscle RING finger, MURF-1
Catalog No. 50-172-0815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.