Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MURF1 Monoclonal Antibody (M316)
GREENER_CHOICE

Catalog No. PIMA547723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547723 Supplier Invitrogen™ Supplier No. MA547723
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a full length human MuRF1 recombinant protein on SDS-PAGE immunoblots.

This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin's N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MURF1
Applications Western Blot
Classification Monoclonal
Clone M316
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene TRIM63
Gene Accession No. Q38HM4, Q91Z63, Q969Q1
Gene Alias E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; iris ring finger protein; Murf; MURF 1; MuRF1; MuRF-1; MURF2; muscle ring finger protein 1; muscle specific ring finger protein 2; muscle-specific RING finger protein 1; OTTHUMP00000008701; RF1; ring finger protein 28; RING-type E3 ubiquitin transferase TRIM63; RNF 28; RNF28; SMRZ; striated muscle RING zinc finger protein; TRIM 63; Trim63; tripartite motif containing 63; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; tripartite motif-containing protein 63
Gene Symbols TRIM63
Host Species Mouse
Immunogen Clone M316 was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal half of human MuRF1. This peptide sequence is highly conserved in rat and mouse MuRF1, and has 50% homology to MuRF2 (TRIM-55).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140939, 433766, 84676
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.