Learn More
AdipoGen Muristerone A (250 µg)
SDP
Supplier:  AdipoGen AGCN20070C250
Encompass_Preferred
Chemical. CAS: 38778-30-2. Formula: C27H44O8. MW: 496.6. Isolated from Ipomoea hederacea seeds. Potent member of the ecdysteroid family. Ecdysone receptor (EcR) agonist. Analog of ecdysone with similar properties to ponasterone A (Prod. No. AG-CN2-0053 http://www.adipogen.com/ag-cn2-0053/ponasterone-a.html ). Inducer of ecdysone-inducible gene expression systems in mammalian cells and transgenic animals. Induces apoptosis in cells transfected with wild-type Bax. Induces expression of beta-galactosidase. Stimulates Bcl-XL mRNA transcription and inhibits TRAIL- and hFasL-induced apoptosis in RKO cells. Insect steroid hormone involved in regulating metamorphosis, causing a response to G2 cell cycle arrest. Major molting hormone in some insects. Has protective effects in plants.
Catalog No. 50-205-3416
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.