Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Muromonab Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541754
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541754 Supplier Invitrogen™ Supplier No. MA541754
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Murine monoclonal antibody specific to CD3 T-cell lymphocyte antigens. More specifically it is a purified murine (mouse) monoclonal antibody, directed against the CD3 (T3) receptor on the surface of human T-cells (T-lymphocytes) cultured using the murine ascites method. Muromonab is 93% monomeric immune globulin G type 2a (IgG2a). Muromonab binds to the T-cell surface glycoprotein CD3 epsilon chain. It appears to kill CD-3 positive cells by inducing Fc mediated apoptosis, antibody mediated cytotoxicity and complement-dependent cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muromonab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 3.95 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias CD3E; OKT3
Host Species Mouse
Immunogen Human CD3E.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.