Learn More
Bachem Muscimol
SDP
Supplier:  Bachem 4004216.0050
Encompass_Preferred
Muscimol, 5-(Aminomethyl)-3(2H)-isoxazolone, 2763-96-4, C4H6N2O2, Mr 114.10, 50mg. Agarin, Pantherine. Muscimol, the major alkaloid in mushrooms of the Amanita family, is a potent, selective agonist of the GABA-A receptor.
Catalog No. 50-260-620
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.