Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Muscimol, GABAA Agonist, >99% Purity, 100 mM/mL

Catalog No. 502704532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mM/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4532 100 mM/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4532 Supplier Abcam Supplier No. AB146673100MM_ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective GABAA agonist with >99% purity.

  • Purity: >99%
  • Potent, selective GABAA agonist
  • Molecular Weight: 114.1 Da
  • Form: Solid
  • Composition: 100% 5-(Aminomethyl)isoxazol-3-ol hydrate
  • Solubility: Soluble in 1 mL water to give specified mM/mL concentration
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule GABAA Receptor
Shipping Condition Ambient
Form Solid
Buffer 100% 5-(Aminomethyl)isoxazol-3-ol hydrate
Concentration 100 mM
Content And Storage Store at 4°C.
Quantity 100 mM/mL
For Use With (Application) GABAA receptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.