Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscle Satellite Cells, Rat anti-Mouse, Clone: SM-C-2.6 , Millipore Sigma™

Catalog No. MABT85725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT85725UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT85725UG Supplier MilliporeSigma Supplier No. MABT85725UG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Muscle Satellite Cells, clone SM C/2.6, Cat. No. MABT857, is a mouse monoclonal antibody that detects muscle satellite cells and has been tested for use in Flow Cytometry, Immunofluorescence, Immunohistochemistry, and Fluorescence Activated Cell Sorting (FACS).

Skeletal muscle satellite cells are quiescent mononucleated myogenic cells that reside between the sarcolemma and basement membrane of terminally-differentiated muscle fibers. They serve as a reserve population of cells with stem-like properties. The number of satellite cells are shown to be highest in postnatal muscle, but declines with age. In their quiescent stage, satellite cells are rounded and mononucleated, and do not express relevant amounts of myogenic proteins. Also, in their quiescent state they are shown to be c-kit-, Sca-1-, CD34+, and CD45-. Satellite cells can proliferate in response to injury and regenerate muscle and produce more satellite cells. Hence, they have been considered to be the only myogenic source for the maintenance of skeletal muscle. When muscle regeneration is initiated, satellite cells changes from their quiescent state to an active stage and begin to proliferative. They can proliferate and then either fuse with each other to form myotubes, which mature into myofibers, or they can fuse with damaged segments of muscle fibers. (Ref.: Fukuda, S et al. (2004). Exp. Cell Res. 296(2); 245-255; Morgan JE and Partridge, TA (2003). Int. J. Biochem. Cell Biol. 35(8); 1151 1156).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscle Satellite Cells
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone SM-C-2.6
Conjugate Unconjugated
Dilution Fluorescence Activated Cell Sorting (FACS) Analysis: A representative lot detected Muscle Satellite Cells in Fluorescence Activated Cell Sorting (FACS) applications (Urbani, L., et. al. (2012). PLoS One. 7(11):e49860). Flow Cytometry Analysis: A representative lot detected Muscle Satellite Cells in Flow Cytometry applications (Ikemoto, M., et. al. (2007). Mol Ther. 15(12):2178-85; Fukada, S., et. al. (2004). Exp Cell Res. 296(2):245-55).Immunohistochemistry Analysis: A representative lot detected Muscle Satellite Cells in Immunohistochemistry applications (Fukada, S., et. al. (2004). Exp Cell Res. 296(2):245-55; Yao, Y., et. al. (2016). Nat Commun. 7:11415).Immunofluorescence Analysis: A representative lot detected Muscle Satellite Cells in Immunofluorescence applications (Fukada, S., et. al. (2004). Exp Cell Res. 296(2):245-55).
Formulation Purified rat monoclonal antibody IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen C2/4 myogenic cells.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone SM/C-2.6 detects quiescent satellite cells from adult mouse muscle.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.