Description
- Allows for the facile and rapid measurement of red blood cells infected with Plasmodium parasites, by identifying red blood cells that stain positive with the nucleic acid reagent provided in the kit
- Minimal sample preparation is required in this no-wash assay to obtain accurate and precise results
Specifications
Specifications
| Quantity | 100 Tests |
| Type | Cell Invasion Assay |