Learn More
Cytiva (Formerly Pall Lab) Mustang Q chromatography membrane in Novasip™ capsule, 10 mL bed volume, 1 in sanitary flange connections
Supplier:  Cytiva CLM05MSTGQP1
Encompass
Disposable Membrane Chromatography Units for Scale-down Process Development, scale-down Mustang membrane unit facilitates process development work. Standard pricing not available. Cytiva requires a new quote with updated pricing for each order.
Catalog No. NC9828755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.