Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mutant HTT Mouse anti-Human,Mouse, Unconjugated, Clone: PHP2, MilliporeSigma™

Catalog No. MABN24502MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN24502MI 25 μL
MABN24501MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN24502MI Supplier MilliporeSigma Supplier No. MABN245025UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mutant HTT Monoclonal antibody specifically detects mutant HTT in Human,Mouse samples. It is validated for Dot Blot, Electron Microscopy, Immunohistochemistry, Inhibits Activity/Function, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mutant HTT
Applications Dot Blot, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone PHP2
Conjugate Unconjugated
Gene Accession No. NP_002102.4
Gene Alias Huntingtin;Huntington disease protein;HD protein
Gene Symbols HTT, HD, IT15
Host Species Mouse
Immunogen GST-tagged recombinant exon1 of wild-type human huntingtin HTTx1 (20Q) protein.
Molecular Weight of Antigen 347.60 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone PHP1 is mouse monoclonal antibody that detects mutated Huntingtin protein. It targets an epitope QAQPLLPQP within the proline-rich domain.
Target Species Human, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.