Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVB12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560468 Supplier Invitrogen™ Supplier No. PA560468
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSTMPEVKDL SEALPETSMD PITGVGVVAS RNRAPTGYDV VAQTADGVDA DLWKDGLFKS KVTRYLCFTR SFSKENSHLG NVL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVB12B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MVB12B
Gene Accession No. Q9H7P6
Gene Alias C9orf28; ESCRT-I complex subunit MVB12B; FAM125B; family with sequence similarity 125, member B; multivesicular body subunit 12B; MVB12B; Protein FAM125B
Gene Symbols MVB12B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MVB12B. Recombinant protein control fragment (Product #RP-100293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.