Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MVP Monoclonal Antibody (LMR5)
GREENER_CHOICE

Catalog No. 01671028
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-028 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-028 Supplier Invitrogen™ Supplier No. MA135590
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LMR5 reacts with an internal epitope of the LRP/Major Vault Protein (P110), which is strongly overexpressed in various human non-P-glycoprotein MDR tumor cell lines.

This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MVP
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LMR5
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation Tissue culture supernatant with 1% BSA and 0.1% sodium azide
Gene MVP
Gene Accession No. Q14764
Gene Alias 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP
Gene Symbols MVP
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9961
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.