Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MW Series Utility Cart with Wire Shelves

Catalog No. 19138770 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W x H):
24 x 18 x 38 in.
30 x 18 x 38 in.
36 x 18 x 38 in.
Handle Material:
Chrome
Stainless Steel
Shelf Material:
Chrome
Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Handle Material Shelf Material
19-138-770 24 x 18 x 38 in. Chrome Chrome
19-138-773 36 x 18 x 38 in. Chrome Chrome
19-138-772 30 x 18 x 38 in. Stainless Steel Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-138-770 Supplier Metro™ Supplier No. MW701
Add to Cart
Add to Cart

Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.

  • Cart load capacity 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
  • One-piece stainless-steel handles add rigidity and durability
  • 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
  • Three tiers
  • Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Product Type Utility Cart
Capacity (Metric) 170 kg
Certifications/Compliance NSF
Color Silver
Description 3-tier Model
Dimensions (L x W x H) 24 x 18 x 38 in.
For Use With (Application) Transporting Materials
Handle Material Chrome
Height (English) 38 in.
Height (Metric) 97 cm
Includes Super Erecta™ 3 Chrome-plated wire shelves with plastic split sleeves, 2 one-piece Chrome-plated handle, 4 in. dia. caster, 3.5 in. donut bumpers
Length (English) 24 in.
Length (Metric) 61 cm
Load Bearing Capacity (English) 375 lb.
Load Bearing Capacity (Metric) 170 kg
Material Steel
No. of Shelves 3
Shelf Material Chrome
Width (English) 18 in.
Width (Metric) 46 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.