Learn More
TARGETMOL CHEMICALS INC MX69 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T365310MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MX69 is the MDM2/XIAP inhibitor blocking the MDM2 protein-XIAP RNA interaction leading to MDM2 degradation. purity: 99%
Catalog No. 50-257-6895
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.