Learn More
MilliporeSigma™ Supelco™ MXP High Pressure Valves for HPLC, PEEK
Catalog No. 111006044 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-6044 1/Pk.
1 options
Catalog No. 11-100-6044 Supplier MilliporeSigma™ Supelco™ Supplier No. 51356U
Add to Cart
Add to Cart
The MXP7900-000 valve is the standard 2-position, 6-port switching valve designed for traditional HPLC and related techniques, used for traditional sample injections or to provide dual-column functionality in your system. The MXP9900-000 valve should be used when the application requires biocompatability.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description MXP9900-000 2-position, 6-port
Material PEEK