Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MXRA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142517 Supplier Thermo Scientific Supplier No. PA5142517
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

MXRA5 Polyclonal Antibody

MXRA5 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MXRA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MXRA5
Gene Accession No. Q9NR99
Gene Alias Adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan; adlican; matrix remodeling associated 5; matrix-remodeling-associated protein 5; matrix-remodelling associated 5; MXRA5
Gene Symbols MXRA5
Host Species Goat
Immunogen Peptide with sequence C-QAETTTLDQDHLET.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25878
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.