Learn More
MilliporeSigma™ Supelco™ MXT Valves for Fast Chromatography, Stainless Steel
Catalog No. 111005989 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-5989 1/Pk.
1 options
Catalog No. 11-100-5989 Supplier MilliporeSigma™ Supelco™ Supplier No. 51343U
You must be signed in and have a business account to purchase online.
The MXT715-000 2-position, 6-port switching valve is ideal for use as a two-column switching valve, enabling the same system to be used with more than one column to easily accommodate multiple users and applications. It can also be used in a traditional injection valve configuration and offers the Rheodyne patented MMB (Make-Before Break) feature for improved reproducibility and system stability.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description MXT715-000 2-position, 6-port
Material Stainless Steel