Learn More
ATCC MYA-1 (CRL-2417) T Lymphoblast Cell Line, Felis catus (Cat) - Suspension Culture
Supplier: ATCC CRL2417
MYA-1 (CRL-2417) is a T lymphoblast cell line derived from peripheral blood of a specific-pathogen-free cat. The line grows in suspension and requires interleukin-2 for proliferation. It is supplied cryopreserved and is intended for research applications under appropriate laboratory conditions.
- Organism: cat (Felis catus).
- Cell type: T lymphoblast.
- Tissue source: peripheral blood.
- Morphology: lymphoblast.
- Growth properties: suspension culture.
- Recommended medium and supplements: RPMI 1640 with 2 mM L-glutamine, 1.5 g/L sodium bicarbonate, 4.5 g/L glucose, 10 mM HEPES, 1.0 mM sodium pyruvate, 0.05 mM 2-mercaptoethanol, 100 units/mL recombinant human IL-2, and 10% fetal bovine serum.
- Incubation conditions: 37°C in a 5% CO2 atmosphere.
- Supply format: standard single vial per unit; exact cell count per vial not specified on manufacturer page.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.