Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYBBP1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596287 Supplier Invitrogen™ Supplier No. PA596287
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLTVLLGVLQ GQQQSLQQGA HSTGSSRLHD LYWQAMKTLG VQRPKLEKKD AKEIPSATQS PISKKRKKKG FLPETKKRKK RKSEDGTPAE DGTPAATGGS QPPSMGRKKR NRTKAKVPAQ ANGTPTTKSP APGAPTRSPS TPAKSPKLQK KNQKPSQVNG APGSPTEPAG QKQHQKALPK KGVLGKSPLS ALARKKARLS LVIRSPSLLQ SGAKKKAQVR KAGKP; Positive Samples: HeLa, SW620, MCF7, Mouse liver, Rat liver; Cellular Location: Cytoplasm, Nucleus, nucleolus.

The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predominantly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYBBP1A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.60 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYBBP1A
Gene Accession No. O35821, Q7TPV4, Q9BQG0
Gene Alias AL024407; AU019902; DBP; MYB binding protein 1a; MYB binding protein (P160) 1a; MYB binding protein 1a; myb-binding protein 1A; Myb-binding protein of 160 kDa; MYBBP1A; nuclear protein P160; P160; p160MBP; p53-activated protein-2; p67MBP; PAP2; PAR interacting protein; PAR-interacting protein; PIP
Gene Symbols MYBBP1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1104-1328 of human MYBBP1A (NP_0553352).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10514, 18432, 60571
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.