Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYBPC2 Monoclonal Antibody (OTI2E1), TrueMAB™, OriGene
SDP

Catalog No. 501683902
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3902 Supplier OriGene Supplier No. TA811203
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the fast-type isoform designated MYBPC2. Both the fast-type and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. MYBPC2 is typically required for strong contractions and functions under anaerobic conditions. It is more similar to the cardiac isoform (MYBPC3) than to the slow-type isoform (MYBPC1) in terms of pCa50-indexed force development, length-independent cooperativity and length dependent activation. It has been suggested that in cardiac and fast muscle MYBPC2 contributes to an internal load, possibly by binding to actin via its N-terminal region.

Specifications

Antigen MYBPC2
Applications Western Blot
Classification Monoclonal
Clone OTI2E1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MYBPC2
Gene Accession No. Q14324
Gene Alias MYBPC, MYBPCF
Gene Symbols MYBPC2
Host Species Mouse
Immunogen Full length human recombit protein of human MYBPC2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4606
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.