Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components
SDP

Catalog No. 22122325
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
NaOH/NALC 2.5% Bulk
NaOH/NALC 3.0% Bulk
Neutralization Buffer B
Resuspension Buffer C
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-122-325 NaOH/NALC 2.5% Bulk
22-122-328 Neutralization Buffer B
22-122-326 NaOH/NALC 3.0% Bulk
22-122-327 Resuspension Buffer C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-122-325 Supplier IMMY Supplier No. TBNN10102.5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Empowers labs to maximize the retention of viable Mycobacteria via rigid pH control throughout the digestion/decontamination process

  • Culture results obtained faster
  • Fewer specimens to reprocess
  • Reduced risk of cross-contamination
  • Easy to process specimens containing blood
  • Reduces sample splitting workload
  • Eliminates user variability, reducing the chances for technical error and allowing samples to be easily processed
  • Advanced buffer reduces the number of samples that need to be split
  • Cleaner smears
  • Visual result confirmation

Specifications

Type NaOH/NALC 2.5% Bulk
For Use With (Application) Mycobacterial testing
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.