Learn More
IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components
Description
- Culture results obtained faster
- Fewer specimens to reprocess
- Reduced risk of cross-contamination
- Easy to process specimens containing blood
- Reduces sample splitting workload
- Eliminates user variability, reducing the chances for technical error and allowing samples to be easily processed
- Advanced buffer reduces the number of samples that need to be split
- Cleaner smears
- Visual result confirmation
Specifications
Specifications
| Type | NaOH/NALC 3.0% Bulk |
| For Use With (Application) | Mycobacterial testing |
| Quantity | 10 Pk. |