Learn More
IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components
SDP
Catalog No. 22122327
Click to view available options
Type:
NaOH/NALC 2.5% Bulk
NaOH/NALC 3.0% Bulk
Neutralization Buffer B
Resuspension Buffer C
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-122-327 Resuspension Buffer C
22-122-328 Neutralization Buffer B
22-122-326 NaOH/NALC 3.0% Bulk
22-122-325 NaOH/NALC 2.5% Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-122-327 Supplier IMMY Supplier No. TBRB3060
Add to Cart
Add to Cart
This item is not returnable. View return policy
Empowers labs to maximize the retention of viable Mycobacteria via rigid pH control throughout the digestion/decontamination process
  • Culture results obtained faster
  • Fewer specimens to reprocess
  • Reduced risk of cross-contamination
  • Easy to process specimens containing blood
  • Reduces sample splitting workload
  • Eliminates user variability, reducing the chances for technical error and allowing samples to be easily processed
  • Advanced buffer reduces the number of samples that need to be split
  • Cleaner smears
  • Visual result confirmation
Specifications
Type Resuspension Buffer C
For Use With (Application) Mycobacterial testing
Quantity 60 x 3 mL