Learn More
Invitrogen™ Mycobacteria LAM Recombinant Rabbit Monoclonal Antibody (5179)
Shop All Thermo Scientific Products

Description
Reactive with: Mycobacterium tuberculosis (6 strains), M. bovis, M. smegmatis, M. leprae, M. avium, M. phlei & M. parafortuitum.
Specifications
Specifications
| Antigen | Mycobacteria LAM |
| Applications | ELISA, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 5179 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Rabbit |
| Purification Method | Protein A |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.