Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycobacterium tuberculosis 16 kDa Monoclonal Antibody (G06I)
GREENER_CHOICE

Catalog No. MA17065
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17065 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17065 Supplier Invitrogen™ Supplier No. MA17065
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7065 detects the Mycobacterium tuberculosis. This antibody recognizes the 16kDa antigen (HspX) of Mycobacterium tuberculosis and M. bovis. MA1-7065 has been successfully used in ELISA and Western blot procedures. The MA1-7065 immunogen is M. tuberculosis lysate.

Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium tuberculosis 16 kDa
Applications ELISA, Western Blot
Classification Monoclonal
Clone G06I
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias m tuberculosis; M. tuberculosis
Host Species Mouse
Immunogen M. tuberculosis lysate.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.