Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Mycobacterium Tuberculosis
SCI_ED

Catalog No. S30184 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30184 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30184 Supplier Eisco™ Supplier No. BS18073
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of mycobacterium tuberculosis, wholemount. Mycobacterium tuberculosis is a gram positive pathogenic bacteria causing tuberculosis in humans. Ideal for microbiology studies at home or in the classroom.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characterisic morphology

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Mounting Medium
Type Microbiology slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Mycobacterium Tuberculosis
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.