Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis Antibody (BGN/1209/3875), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10062769G
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062769G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062769G Supplier R&D Systems Supplier No. NB10062769G

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mycobacterium tuberculosis Monoclonal antibody specifically detects Mycobacterium tuberculosis in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Mycobacterium tuberculosis
Applications Western Blot, ELISA
Classification Monoclonal
Clone BGN/1209/3875
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias M. tuberculosis
Host Species Mouse
Immunogen Recombinant 38 kDa antigen from Mycobacterium tuberculosis.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.