Learn More
Description
PA1-7231 detects the Mycobacterium tuberculosis. PA1-7231 has been successfully used in immunohistochemistry procedures. The PA1-7231 immunogen is tuberculosis PPD (purified protein derivative). Store at 4ºC for up to 6 months. For longer storage, store at -20ºC, Avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | Mycobacterium tuberculosis |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | m tuberculosis; M. tuberculosis |
| Host Species | Rabbit |
| Immunogen | Purified PPD. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
