Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycophenolic acid, 98%

Catalog No. AAJ6190506
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6190506 5 g
AAJ6190503 1 g
AAJ6190509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6190506 Supplier Thermo Scientific Chemicals Supplier No. J6190506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24280-93-1 | C17H20O6 | 320.341 g/mol

It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24280-93-1
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.341
MDL Number MFCD00036814
InChI Key HPNSFSBZBAHARI-RUDMXATFSA-N
Synonym mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid
PubChem CID 446541
ChEBI CHEBI:168396
IUPAC Name (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC

Specifications

Melting Point 139°C to 141°C
Color White
Quantity 5 g
Beilstein 1295848
Merck Index 14,6327
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 320.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mycophenolic acid
Hazard Category H302-H341-H360D-H372
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 246-119-3
RTECSNumber MP8050000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.