CAS RN 24280-93-1
Mycophenolic acid, 98%
CAS: 24280-93-1 Molecular Formula: C17H20O6 Molecular Weight (g/mol): 320.341 MDL Number: MFCD00036814 InChI Key: HPNSFSBZBAHARI-RUDMXATFSA-N Synonym: mycophenolic acid,mycophenolate,melbex,myfortic,mycophenolsaeure,acido micofenolico,micofenolico acido,acide mycophenolique,acidum mycophenolicum,6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid PubChem CID: 446541 ChEBI: CHEBI:168396 IUPAC Name: (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid SMILES: CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC
Thermo Scientific Chemicals Mycophenolic acid, 98%
CAS: 24280-93-1 Molecular Formula: C17H20O6 Molecular Weight (g/mol): 320.34 InChI Key: HPNSFSBZBAHARI-RUDMXATFSA-N Synonym: mycophenolic acid,mycophenolate,melbex,myfortic,mycophenolsaeure,acido micofenolico,micofenolico acido,acide mycophenolique,acidum mycophenolicum,6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid PubChem CID: 446541 ChEBI: CHEBI:168396 IUPAC Name: (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid SMILES: CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC
Mycophenolic Acid, Spectrum™ Chemical
CAS: 24280-93-1 Molecular Formula: C17H20O6 Molecular Weight (g/mol): 320.34 InChI Key: HPNSFSBZBAHARI-RUDMXATFSA-N IUPAC Name: (4E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydro-2-benzofuran-5-yl)-4-methylhex-4-enoic acid SMILES: COC1=C(C)C2=C(C(=O)OC2)C(O)=C1C\C=C(/C)CCC(O)=O
MilliporeSigma™ Calbiochem™ Mycophenolic Acid
An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of inosine monophosphate dehydrogenase and prevents the formation of XMP and GMP.
Mycophenolic Acid, MP Biomedicals
CAS: 24280-93-1 Molecular Formula: C17H20O6 Molecular Weight (g/mol): 320.341 MDL Number: MFCD00036814 InChI Key: HPNSFSBZBAHARI-RUDMXATFSA-N Synonym: mycophenolic acid,mycophenolate,melbex,myfortic,mycophenolsaeure,acido micofenolico,micofenolico acido,acide mycophenolique,acidum mycophenolicum,6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid PubChem CID: 446541 ChEBI: CHEBI:168396 IUPAC Name: (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid SMILES: CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC
Mycophenolic Acid 98.0+%, TCI America™
CAS: 24280-93-1 Molecular Formula: C17H20O6 Molecular Weight (g/mol): 320.341 MDL Number: MFCD00036814 InChI Key: HPNSFSBZBAHARI-RUDMXATFSA-N Synonym: mycophenolic acid,mycophenolate,melbex,myfortic,mycophenolsaeure,acido micofenolico,micofenolico acido,acide mycophenolique,acidum mycophenolicum,6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid PubChem CID: 446541 ChEBI: CHEBI:168396 IUPAC Name: (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid SMILES: CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC
R&D Systems™ Mycophenolic acid
CAS: 24280-93-1 Molecular Formula: C17H20O6