Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-168.5), Invitrogen™

Catalog No. p-7245238
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546110 100 μg
01-671-823 500 μg
PIMA546157 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546110 Supplier Invitrogen™ Supplier No. MA546110
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10793 detects Mycoplasma bovis. MA1-10793 has been successfully used in ELISA applications. The MA1-10793 immunogen is Mycoplasma bovis cells.

This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma bovis
Applications ELISA
Classification Monoclonal
Clone MY10-168.5
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias m bovis; M. bovis; mycoplasmen
Host Species Mouse
Immunogen Mycoplasma bovis cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.