Learn More
Enzo Life Sciences MyD88 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADICSA510E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human MyD88. The sequence is completely conserved in mouse. Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.