Learn More
CPC Scientific H-Val-His-Phe-Phe-Lys-Asn-Ile-Val-Thr-Pro-Arg-Thr-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MBP001A
Encompass
This peptide is the immunodominant eptitope of Myelin Basic Protein, which has been shown to induce the proliferation of an encephalitogenic, myelin basic protein-specific T cell line. Active immunization of rats with this peptide induced acute experimental autoimmune encephalomyelitis. This protein region is highly conserved among mammalian species.ONE-LETTER SEQUENCE: VHFFKNIVTPRTPMOLECULAR FORMULA: C74H114N20O17MOLECULAR WEIGHT:1555.84STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [118506-26-6], RESEARCH AREA: Multiple SclerosisREFERENCES: R.E.Jones et al., J. Neuroimmunol., 37, 203 (1992)
Catalog No. 50-210-9705
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.