Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Myelin Basic Protein Peptide, Mouse, Synthetic, >90% Purity, 1 mg

Catalog No. 502710963
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0963 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0963 Supplier Abcam Supplier No. AB403891MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide for use in blocking applications with >90% purity.

  • Purity: >90%
  • Form: Liquid
  • Applications: BL
  • Formulation: pH 6.75; includes 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, and 0.001% Sorbitan monolaurate, ethoxylated
  • Type: Synthetic
  • Species: Mouse
  • Citation Count: 1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Peptide-based Substrate
Target Molecule Myelin Basic Protein (Mouse)
Reactivity Mouse
Shipping Condition Blue Ice
Label or Dye Unconjugated
Buffer 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75
Concentration 1 mg/mL
Content And Storage Store at -20°C.
Form Liquid
Formulation pH 6.75, 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
Quantity 1 mg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.