Learn More
CPC Scientific H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Pro-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MOG002A
Encompass
In contrast to the encephalitogenic rat homolog, this peptide is not a major T-cell epitope in multiple sclerosis propably due to its lower amount of helical structure formed when dissolved in micellar sodium dodecyl sulfate. This peptide induced anti-MOG(35-55) autoantibody production in C57BL/6 mice.ONE-LETTER SEQUENCE: MEVGWYRPPFSRVVHLYRNGKMOLECULAR FORMULA: C120H179N35O28S1MOLECULAR WEIGHT:2592.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [163158-19-8], RESEARCH AREA: Multiple SclerosisREFERENCES: N.Kerlero de Rosbo et al., Eur. J. Immunol., 27, 3059 (1997); S.Albouz-Abo et al., Eur. J. Biochem., 246, 59 (1997); M.F.Mesleh et al., Neurobiol. Dis., 9, 160 (2002); Oliver, A. J. Immunology 171, 462 (2003); Stromnes, I. et al. Nature protocols 1, 1810 (2006)
Catalog No. 50-210-9815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.