Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
BM-4 recognizes a nuclear antigen expressed in human granulocytes (83%) monocytes (20%) and myeloid precursor cells residing in lymphoid and non-lymphoid tissues. BM-4 is an early marker of myeloid differentiation. It also reacts with a subset of myeloid leukemia cells. BM-4 has no reactivity with any other cell type in human tissues.
This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
Specifications
Specifications
| Antigen | Myeloid Specific Antigen |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | BM-4 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MNDA |
| Gene Accession No. | P41218 |
| Gene Alias | Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
