Learn More
Diagnostic Biosystems MYELOPEROXIDASE Myeloperoxidase
Supplier:  Diagnostic Biosystems RP05305
Encompass_Preferred
This antibody reacts with human myeloperoxidase. It stains granules of neutrophils, granulocytesin spleen, bone marrow, tonsil and blood smears.
Catalog No. 50-238-5706
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.