Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Myeloperoxidase/MPO Activity Assay Kit (Fluorometric)
SDP

Catalog No. NB340273
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340273 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340273 Supplier Novus Biologicals™ Supplier No. NBP324524

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Assay Kit (Fluorometric)

Specifications

Content And Storage Storage of components varies. See protocol for specific instructions.
Gene ID (Entrez) 4353
Gene Symbol MPO
Quantity 96 Tests
Synonym EC 1.11.1, EC 1.11.1.7, myeloperoxidase
Research Category Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
Species Non-species specific
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.