Learn More
Medchemexpress LLC 1-(3-methyl-3-((3-(trifluoromethyl)phenyl)amino)pyrrolidin-1-yl)prop-2-en-1-one | 2416417-65-5 | 99.4% | C15H17F3N2O | 10MG
Supplier: Medchemexpress LLC HY13960310MG
MYF-01-37 is a small-molecule covalent inhibitor of TEAD that targets cysteine 380 and produces reversible inhibition of the YAP-TEAD protein-protein interaction. It is characterized for research use with defined molecular properties and high analytical purity.
- Targets cysteine 380 on TEAD, enabling covalent engagement.
- Inhibits YAP-TEAD interaction relevant to Hippo/Wnt signaling pathways.
- High purity for reproducible experimental results.
- Available as solid and as DMSO solution for flexible assay setup.
- Intended for research use only; not for clinical or diagnostic use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.