Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518927
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518927 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518927 Supplier Invitrogen™ Supplier No. PA518927
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Myogenic factor 5 (MYF5) is a gene located on chromosome 12q21.31, encoding a basic helix-loop-helix (bHLH) transcription factor essential for skeletal muscle development and differentiation. MYF5 is one of the key members of the myogenic regulatory factor (MRF) family, which also includes MyoD, myogenin, and MRF4. It plays a crucial role in myogenesis by regulating the expression of genes necessary for muscle precursor cell proliferation and differentiation into mature muscle fibers. During embryonic development, MYF5 is involved in the formation of muscle tissue and maintenance of muscle progenitor cells. Mutations or dysregulation in MYF5 can lead to developmental abnormalities in muscle formation and are implicated in muscle-related disorders. Research into MYF5 continues to explore its regulatory mechanisms in muscle biology and potential therapeutic applications in muscular dystrophies and muscle repair processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYF5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MYF5
Gene Accession No. P13349
Gene Alias B130010J22Rik; basic helix-loop-helix nuclear protein; bHLHc2; cb641; Class C basic helix-loop-helix protein 2; muscle regulatory protein Myf5; MYF5; myf-5; myf5 protein; MyoD family; myogenic factor 5; myogenic factor-5; myogenic regulatory factor 5; myogenic regulatory factor MYF-5; unnamed protein product; zgc:110509
Gene Symbols MYF5
Host Species Goat
Immunogen Synthetic peptide sequence (PECNSPVWSRKSST) corresponding to the internal amino acids of MYF5 (aa 168-182).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.