Learn More
Invitrogen™ MYF6 Polyclonal Antibody

Description
MYF6 Polyclonal Antibody detects endogenous levels of MYF6 protein.
Specifications
Specifications
| Antigen | MYF6 |
| Applications | Immunohistochemistry, Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Myf6 |
| Gene Accession No. | P15375, P19335, P23409 |
| Gene Alias | bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6 |
| Gene Symbols | Myf6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.