Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH10 Monoclonal Antibody (GT246)
GREENER_CHOICE

Catalog No. PIMA527767 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527767 Supplier Invitrogen™ Supplier No. MA527767
  • $640.00
    $506.65 / Each of 1

    Save $133.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Positive Control: HeLa, MYH10-transfected 293T, NIH-3T3, mouse brain Predicted Reactivity: Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MYH10 encodes a member of the myosin superfamily. The protein represents a conventional non-muscle myosin; it should not be confused with the unconventional myosin-10 (MYO10). Myosins are actin-dependent motor proteins with diverse functions including regulation of cytokinesis, cell motility, and cell polarity. Mutations in this gene have been associated with May-Hegglin anomaly and developmental defects in brain and heart. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH10
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT246
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MYH10
Gene Accession No. P35580, Q61879, Q9JLT0
Gene Alias 5730504C04Rik; 9330167F11Rik; cellular myosin heavy chain, type B; flectin; Fltn; MCH-B; MCH-B(B2); mKIAA3005; MYH10; Myhn2; Myhn-2; Myosin heavy chain 10; myosin heavy chain 10, non-muscle; myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy chain 10, non-muscle; myosin, heavy polypeptide 10, non-muscle; myosin-10; NMHC II-B; NMHC-B; NMHCII-B; NMMHC IIB; NMMHC II-b; NMMHCB; NMMHC-B; NMMHC-IIB; nonmuscle myosin; Non-muscle myosin heavy chain B; nonmuscle myosin heavy chain IIB; non-muscle myosin heavy chain IIb; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799; SMemb
Gene Symbols MYH10
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human MYH10. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4628, 77579, 79433
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.